The shelf life is 2 years for all consumables kept in the recommended storage conditions (temperatures from 5 to 30°C; humidity between 10 and 80%). Contact your local representative (https://www.sigmaaldrich.com/collections/offices) for more information.
CDUFBC001
Biopak® C Clinical Cartridge
Removes bacteria and bacterial by-products from water. For use with AFS® and Milli-Q® CLX systems.
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
sterility
0.22 μm filtered
feature
better assay reliability, Reduces the need for frequent analyzer calibration
composition
(polysulfone hollow fibers)
packaging
pkg of 1 unit
manufacturer/tradename
BioPak®
parameter
≥1 bar pressure (3 bar)
0.5 mL/min air diffusion at 2 bar
1.5 L/min flow rate at 1 bar and 20 °C
5 °C temp. range (35 °C)
volume
100 L
100000 mL
total organic carbon (TOC) residue
<500 ppb (after 5 L Rinsing)
pore size
0.22 μm pore size
fitting
1/4 in. inlet connection (Gaz (Straight) Luer Slip F)
8 mm outlet connection (PTFE)
compatibility
for use with AFS®
for use with Milli-Q® CLX
shipped in
ambient
Related Categories
General description
The cartridge efficiently removes bacteria and bacterial by-products, such as alkaline phosphatase (ALP), from analyzer feed water, which is used to dilute reagents, make blanks, and rinse tubing and probes. This results in a more stable baseline and better assay reliability.
The Biopak® C ultrafiltration cartridge is composed of polysulfone hollow fibers. This membrane optimizes the rejection of bacterial ALP and bacteria while maintaining a high flow rate.
Application
Features and Benefits
- Produces ALP-free water (< 1 μUnits/μL).
- Ensures bacteria levels below 10 cfu/mL (typically < 1 cfu/mL).
- Designed to remove endotoxins, and to perform a pyrogen log reduction value superior or equal to 4, for pyrogen feed concentration between 1000 and 10 000 EU/mL, and a nuclease reduction from 1 μg/mL to less than 0.01 ng/mL at 1 bar feed water pressure.
- Supports stable assay baselines, resulting in better assay reliability.
- Reduces the need for frequent analyzer calibration.
- Eliminates the need for frequent analyzer decontamination that results in costly downtime.
- Includes: Self-adhesive label.
- Replaced every 6 months for optimal results.
Components
Other Notes
- Organism Retention: Bacteria and pyrogens
- Mode of Action: Ultrafiltration
- Application: Cell culture media, molecular biology
- Intended Use: Bioburden reduction
- Instructions for Use: This item provides water filtered through ultrafiltration hollow fibers. Refer to insert, section "replacement" in the product package.
- Storage Statement: Store in a dry location.
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the shelf life for Filter?
1 answer-
Helpful?
-
-
How are consumables changed?
1 answer-
To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.
Helpful?
-
-
How can I know which consumables can be used with my system?
1 answer-
To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.
Helpful?
-
-
What steps are you taking towards improved sustainability?
1 answer-
Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.
Helpful?
-
-
Can I arrange to have my consumables automatically ordered and delivered once a year?
1 answer-
It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).
Helpful?
-
-
Can consumables be opened and refilled?
1 answer-
Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.
Helpful?
-
-
If I have a final filter that I use only occasionally (e.g., Biopak®) and have another filter (e.g. Millipak®) that I most of the time, can I keep the 'occasional-use' filter for use again at a later time?
1 answer-
It is not possible to re-use a filter that has been used and then removed because air intake during replacement creates an airlock and blocks water passing through the filter. If you regularly need two different filters, we recommend using two POD dispensers.
Helpful?
-
-
How frequently do the various consumables need to be changed?
1 answer-
The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.
Helpful?
-
-
What is the consumables delivery time?
1 answer-
Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.
Helpful?
-
-
Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?
1 answer-
Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service