CCW4115-50X2ML
Yeast Extract Broth
ready-to-use, tube of 50 × 2 mL, liquid, nonselective for Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
EP 2.6.1
JP 4.06
USP 71
form
liquid
shelf life
9 mo.
feature
ready-to-use
packaging
tube of 50 × 2 mL
technique(s)
microbiological culture: suitable
application(s)
cosmetics
food and beverages
pharmaceutical
components
: 6 g/L (Tryptone )
: 3 g/L (Yeast )
shipped in
2-8°C
storage temp.
2-8°C
suitability
nonselective for Media
Related Categories
General description
Yeast Extract Broth is a water-soluble, peptone-like derivative obtained from yeast (Saccharomyces) cells. It is prepared through the proteolytic autolysis of the cells, with the autolytic activity being arrested by a heating process. The controlled autolysis ensures the preservation of naturally occurring B-complex vitamins.
Yeast Extract is a versatile, growth-enhancing nutritive substrate that can replace serum in cell cultures. And, it maintains an endotoxin value of less than 500 EU/g.
Yeast Extract is a versatile, growth-enhancing nutritive substrate that can replace serum in cell cultures. And, it maintains an endotoxin value of less than 500 EU/g.
Application
Yeast Extract Broth is used to detect microorganisms from water samples.
Analysis Note
- Utilize the membrane filtration method and impregnate a sterile pad with the medium
- Deposit the membrane filter on the pad and ensure protection against dehydration
- Adjust temperature and incubation time as per internal regulations or laboratory methodologies
- Collect, dilute, and prepare samples following specifications, directives, official standard regulations, or expected results
- Incubate aerobically at 36±2ºC for 44±4h
- Note that different incubation times or temperatures may be required based on the sample or normative ISO factors
- Enumerate all colonies on the membrane surface post-incubation
- Recognize the significant impact of the material and quality of the membrane filter on microorganism recovery
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service