Skip to Content
Merck
All Photos(1)

Key Documents

CBL305

Sigma-Aldrich

Anti-Serum Amyloid P Antibody, clone 5.4D.3B

clone 5.4D.3B, Chemicon®, from mouse

Synonym(s):

Component P

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
£445.00

£445.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
£445.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

£445.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

5.4D.3B, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... APCS(325)

General description

Amyloid consists mainly of rigid, non-branching protein fibrils, together with rod-like aggregates of a pentagonal-shaped glycoprotein called amyloid P protein. Amyloid P protein, also known as component P, comprises 10 per cent of amyloid tissue and is present in all but the central nervous system forms of amyloid. Amyloid P protein is a constituent of normal basement membranes and the microfibrillaryelastic fiber network.

Application

Anti-Serum Amyloid P Antibody, clone 5.4D.3B is an antibody against Serum Amyloid P for use in ELISA.
Development of ELISA assay for the estimation of human serum amyloid P using conjugated CBL304.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service