Skip to Content
Merck
All Photos(1)

Key Documents

AX1698A

Supelco

Composite 5

Original Formulation AQUASTAR®

Synonym(s):

Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

form

liquid

transition temp

flash point 46.1 °C

shipped in

ambient

storage temp.

room temp

Application

AX1698A-6, AX1698A-3 and AX1698A-5 are packaged in 38/430 poly coated glass bottles.

Analysis Note

Color: Red-brown
Form: Liquid
Titer (mg water per mL): 5.0mg/mL min

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Oral - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus, Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

115.0 °F

Flash Point(C)

46.1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service