Skip to Content
Merck
All Photos(3)

Key Documents

ASTM03701

Millipore

ASTM® Color Standards

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£205.00

£205.00


Estimated to ship on09 April 2025Details



Select a Size

Change View
1 EA
£205.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NB.24

£205.00


Estimated to ship on09 April 2025Details


manufacturer/tradename

ASTM®

Quality Level

shipped in

ambient

General description

For use in color rating filter membranes for aviation turbine fuels in accordance
with ASTM Test Method D2276.

For use with the Fluid Sampling Kit.

Applications:
ASTM Test Method D2276

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service