Skip to Content
Merck
All Photos(1)

Key Documents

AG546

Sigma-Aldrich

β amyloid 11-40, rat and mouse

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
£1,450.00

£1,450.00


Estimated to ship on05 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 μG
£1,450.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

£1,450.00


Estimated to ship on05 March 2025Details


Request a Bulk Order

biological source

rat

Quality Level

Assay

>95%

form

powder

mol wt

Mw 3170.74 Da

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

UniProt accession no.

shipped in

wet ice

Gene Information

mouse ... APP(11820)
rat ... APP(54226)

General description

Glu-Val-Arg-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

Comparison of Rat and Human Beta-Amyloid (11-40) sequence
EVRHQKLVFFAEDVGSNKGAIIGLMVGGVV - Rat, MW = 3170.74
EVHHQKLVFFAEDVGSNKGAIIGLMVGGVV - Human, MW = 3151.7
Product Source: E. coli

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

White lyophilized powder. Resuspend in 1% NH4OH at a concentration of 1 mg/mL. Sonicate for 30 seconds to 1 minute after it has gone into solution. To bring into your solution: After resuspension, add 5x or 10x buffer stock (PBS, TBS) and water to bring to 1x buffer.

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C to -70°C for up to 2 weeks in undiluted aliquots. Avoid freeze/thaw cycles to avoid aggregation.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service