Skip to Content
Merck
All Photos(1)

Key Documents

AG316

Sigma-Aldrich

Acetylcholine Receptor-ε, control peptide for AB5938

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.21

concentration

1.0 mg/mL

Quality Level

technique(s)

cell based assay: suitable

UniProt accession no.

Gene Information

mouse ... CHRNE(11448)

General description

FUNCTION: SwissProt: Q04844 # After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
SIZE: 493 amino acids; 54697 Da
SUBUNIT: Pentamer of two alpha chains, and one each of the beta, delta, and gamma (in immature muscle) or epsilon (in mature muscle) chains.
SUBCELLULAR LOCATION: Cell junction, synapse, postsynaptic cell membrane; Multi-pass membrane protein.
DISEASE: SwissProt: Q04844 # The muscle AChR is the major target antigen in the autoimmune disease myasthenia gravis [MIM:254200]. Myasthenia gravis is characterized by sporadic muscular fatigability and weakness, occurring chiefly in muscles innervated by cranial nerves, and characteristically improved by cholinesterase- inhibiting drugs. & Defects in CHRNE are a cause of slow-channel congenital myasthenic syndrome (SCCMS) [MIM:601462, 254200]. Congenital myasthenic syndromes (CMS) are inherited disorders of neuromuscular transmission that stem from mutations in presynaptic, synaptic, or postsynaptic proteins. Postsynaptic disorders result from mutations in proteins forming the subunits of the muscle acetylcholine receptor (AChR). The kinetic abnormalities of AChR result in either prolonged ion channel activations that underlie slow-channel myasthenic syndromes (SCCMS) or abbreviated channel activations that underlie the abnormally rapid decay of endplate currents in fast-channel syndromes (FCCMS). A third disorder associated with postsynaptic CMS is called CMS type Id (CMS1d), and could also result from mutations in the proteins forming the subunits of the muscle AChR. Mutations underlying SCCMS cause a gain of function and usually show dominant inheritance. & Defects in CHRNE are a cause of fast-channel congenital myasthenic syndrome (FCCMS) [MIM:608930]. Mutations underlying FCCMS cause a loss of function and show recessive inheritance. & Defects in CHRNE are a cause of congenital myasthenic syndrome type Id (CMS1d) [MIM:608931]; also called congenital myasthenic syndrome associated with acetylcholine receptor deficiency. Mutations underlying AChR deficiency cause a loss of function and show recessive inheritance.
SIMILARITY: SwissProt: Q04844 ## Belongs to the ligand-gated ionic channel (TC 1.A.9) family.

Application

For use in blocking the reactivity of AB5938.

Optimal working dilution must be determined by the end user.

Physical form

Liquid in PBS.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service