Skip to Content
Merck
All Photos(2)

Key Documents

ABT340

Sigma-Aldrich

Anti-AES Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Amino-terminal enhancer of split, GRG, ESP1, TLE5, Gp130-associated protein GAM, Protein ESP1, Protein GRG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TLE5(166)

General description

Amino-terminal enhancer of split (AES), also known as GRG or ESP1 or TLE5 or Gp130-associated protein GAM, is a member of the Groucho/ transducin-like enhancer of split (TLE) family. AES belongs to a group of transcriptional co-repressors that play an important role in neurogenesis, segmentation and sex determination. AES is suggested to be a dominant repressor towards other family members of the TLE family and might be required for the initiation and maintenance of the differentiated state. AES has been shown to inhibit the NF-kappa-B-regulated gene expression. AES forms a complex with Bit1 (Bcl-2 inhibitor of transcription 1). Recent studies suggest that this complex turns off a survival-promoting gene transcription program controlled by the TLE protein family. The apoptosis of cells transfected with AES and Bit1 implies that the Bit1-AES pathway contributing to anoikis is regulated by integrins, and more specifically – with the Alpha5Beta1 integrin. AES is found predominantly in muscle, heart and placenta. Additionally, in fetal tissues, AES is expressed highly in the heart, lung, kidney, brain and liver.

Specificity

This antibody recognizes the C-terminus of AES.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human AES.

Application

Anti-AES Antibody is a highly specific rabbit polyclonal antibody, that targets Amino-terminal enhancer of split & has been tested in western blotting, ICC & Immunofluorescence.
Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected AES in HeLa cells.

Immunofluorescence Analysis: 20 µg/mL from a representative lot detected AES in HeLa cells.
Research Category
Cell Structure

Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected AES in 15 µg of HEK293 cell lysate.

Target description

~22 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 2 isoforms at ~22 kDa and ~29 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service