Skip to Content
Merck
All Photos(1)

Key Documents

ABT1383

Sigma-Aldrich

Anti-phospho-Keratin-18 Antibody (Ser52)

from rabbit, purified by affinity chromatography

Synonym(s):

Keratin, type I cytoskeletal 18, Ser52-phosphorylated, Cell proliferation-inducing gene 46 protein, Ser52-phosphorylated, CK-18, Ser52-phosphorylated, Cytokeratin-18, Ser52-phosphorylated, K18, Ser52-phosphorylated, Keratin-18, Ser52-phosphorylated

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£274.00

£274.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
£274.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

£274.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer52)

Gene Information

human ... KRT18(3875)

General description

Keratin, type I cytoskeletal 18 (UniProt P05783; also known as Cell proliferation-inducing gene 46 protein, CK-18, Cytokeratin-18, K18, Keratin-18) is encoded by the KRT18 (also known as CYK18, PIG46) gene (Gene ID 3875) in human. Cytokeratin-18 (Keratin-18 or K18) belongs to a family of highly conserved epithelial-specific intermediate filament (IF) proteins that play an important role in the mechanical stability and integrity of epithelial cells and tissues. Each epithelium is characterized by its unique keratin expression pattern. All epithelial cells typically express at least one type-I and one type-II keratin, which form noncovalent heteropolymers. Keratin 18 is a type I intermediate filament protein that pairs with keratin 8 (K8/K18) in simple epithelia and plays an important role in protecting cells from stress, injury, and apoptosis. K18 is the only type-I keratin in adult hepatocytes and the major type-I keratin in many colonic tumor cell lines.The structure and function of K8/18 are regulated by posttranslational modifications, such as glycosylation, ubiquitination, and phosphorylation, including K8 phosphorylation on Ser8, Ser23, Ser73, Ser431, and K18 phosphorylation on Ser33 and Ser52. K18 phosphorylation has been proposed as a progression marker for human chronic hepatitis B (CHB). Progressively higher Ser52 phosphorylation is reported in non-cirrhotic and cirrhotic CHB livers, and progressive increase of Ser33 and Ser52 phosphorylation correlates with the elevation of both histological lesions and enzymatic activities of alanine aminotransferase in non-cirrhotic CHB livers.

Specificity

Target phosphorylation site is not conserved in murine and rat species.
This rabbit polyclonal antibody detected upregulated cellular Keratin-18 Ser52 phosphorylation upon phosphatase inhibitors treatment. The Ser52 numbering is based on keratin-18 sequence without the initiation Met1, which was removed posttranslationally.

Immunogen

KLH-conjugated linear peptide corresponding to human Keratin-18 target region sequence containing phosphorylated Ser52.

Application

This rabbit polyclonal Anti-phospho-Keratin-18 (Ser52) Antibody, Cat. No. ABT1383 is validated for use in Western Blotting for the detection of Keratin-18 Ser52 phosphorylation.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected upregulated Keratin-18 Ser52 phosphorylation in 10 µg lysate from phosphatase inhibitors Calyculin A- and Okadaic acid-treated HEK293 cells.

Target description

~48 kDa observed. 47.93 kDa (Met1 removed) calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service