Skip to Content
Merck
All Photos(2)

Key Documents

ABN648

Sigma-Aldrich

Anti-TRESK Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Potassium channel subfamily K member 18, TWIK-related individual potassium channel, TWIK-related spinal cord potassium channel

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
£295.00

£295.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
£295.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

£295.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Potassium channel subfamily K member 18, also known as TWIK-related individual potassium channel or TWIK-related spinal cord potassium channel, and encoded by the gene name KCNK18 or TRESK or TRIK, is a two-pore domain K+ channel that induces outward rectification and functions as a background K+ channel that is abundantly expressed in the spinal cord, thymus, spleen, and leukemic Tlymphocytes. TRESK is activated by increased cytoplasmic calcium concentration through calcineurin as well as by acetylcholine and histamine. TRESK may play a role in the regulation of T-cell function and could be an excellent target to treat T-cell originated immune dysfunction. Mutations in TRESK have been assoicated with migraine with or without aura 13.

Immunogen

KLH-conjugated synthetic peptide corresponding to the center region of human TRESK.

Application

Anti-TRESK Antibody is an antibody against TRESK for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected TRESK in rat brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected TRESK in rat brain cells.



Quality

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: 2 μg/mL of this antibody detected TRESK in Rat brain tissue lysate.

Target description

~40 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service