Skip to Content
Merck
All Photos(1)

Key Documents

ABN1738

Sigma-Aldrich

Anti-SET-beta Antibody

serum, from rabbit

Synonym(s):

Protein SET-beta, HLA-DR-associated protein II beta, IGAAD beta isoform, Inhibitor of granzyme A-activated DNase beta isoform, PHAPII beta isoform, I-2PP2A beta isoform, Phosphatase 2A inhibitor I2PP2A beta isoform, TAF-I beta, Template-activating factor

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
£290.00

£290.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
£290.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

£290.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SET(6418)

General description

Protein SET (UniProt Q9EQU5; also known as I-2PP2A, Phosphatase 2A inhibitor I2PP2A, TAF-I, Template-activating factor I) is encoded by the Set gene (Gene ID 56086) in murine species. Protein SET (patient SE translocation) adopted its name when it was originally identified in a leukemia study in 1992. SET interacts with signaling pathways that promote normal cell growth and proliferation, as well as tumorigenesis and metastasis. SET protein inhibits PP2A by enhancing lyase activity of P450c17, a substrate for PP2A when it is Ser- and Thr-phosphorylated. Alternative splicings result in multiple SET isoforms, including SET-alpha (TAF-I alpha), SET-beta (TAF-I beta), where the N-terminal 37-amino acid sequence in Set-alpha is replaced with a different sequence of a shorter length in Set-beta. In addition, there exists a shorter spliced Set-beta isoform without the nuclear localization signal (NLS). Both Set-alpha and SET-beta are predominantly localized in the nucleus, but differ in their expression levels among various cell and tissue types. SET-beta is resides predominantly in the brain relative to Set-alpha, whereas the opposite pattern was observed in tissues such as the spleen. SET-beta functions have been described both in the nucleus and cytoplasm. The presence of a nuclear localization signal (NLS) and phosphorylation govern SET-beta isoforms cellular localization. Proteolytic cleavage of SET-beta at Lys176 by granzyme A or at Asn-175 by asparagine endopeptidase results in the release of GzmA-activated DNase (GAAD) from a multiprotein complex. Uninhibited in the absence of the full-length Set-beta, GAAD causes cell death by its DNA-nicking activity.

Specificity

This polyclonal antiserum targets sequence specific to the spliced isoform SET-beta (TAF-I beta) of human, murine, and rat species (UniProt Q01105-2, Q9EQU5-2, Q63945-2). The epitope sequence is not present in SET-alpha (TAF-I alpha) or any other human, murine, and rat protein SET spliced isoforms reported by UniProt.

Immunogen

Epitope: N-terminus
Synthetic peptide corresponding to a sequence near the N-terminus of mouse/rat SET-beta.

Application

Anti-SET-beta, Cat. No. ABN1738, is a highly specific rabbit polyclonal antiserum that targets Protein SET and has been tested in Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected predominant nuclear SET-beta immunoreactivity in 4% paraformaldehyde-fixed, 0.2% Triton X-100-permeabilized P4 rat retinal ganglion cells (RGCs) by fluorescent immunocytochemistry, while distinct cytoplasmic and membrane punctae were detected without permeabilization (Trakhtenberg, E.F., et al. (2014). J. Neurosci. 34(21):7361-7374).

Immunofluorescence Analysis: A representative lot detected predominant nuclear SET-beta immunoreactivity among retinal ganglion cells (RGCs) by fluorescent immunohistochemistry staining of 4% paraformaldehyde-fixed, 0.5% Triton X-100-permeabilized retinal cryosections from E19, P8, and adult rats (Trakhtenberg, E.F., et al. (2014). J. Neurosci. 34(21):7361-7374).

Immunoprecipitation Analysis: A representative lot co-immunoprecipitated PP2A-A with SET-beta from E18 rat hippocampal homogenates (Trakhtenberg, E.F., et al. (2014). J. Neurosci. 34(21):7361-7374).

Western Blotting Analysis: A representative lot detected SET-beta in rat retinal retinal ganglion cell (RGC) lysate and in E18 rat hippocampal homogenates (Trakhtenberg, E.F., et al. (2014). J. Neurosci. 34(21):7361-7374).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antiserum detected SET-beta in 10 µg of Jurkat cell lysate.

Target description

~38 kDa observed. Band size appears larger than the calculated molecular weight of 31.97 kDa (Met1 removed) due to posttranslational modification (acetylation and/or phosphorylation). Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service