Skip to Content
Merck
All Photos(1)

Key Documents

ABE2865

Sigma-Aldrich

Anti-Meis1/2

from rabbit

Synonym(s):

Homeobox protein Meis1, Myeloid ecotropic viral integration site 1, Homeobox protein Meis2, Meis1-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

salamander (based on 100% sequence homology), human (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
immunofluorescence: suitable
western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Meis2(17536)

General description

Homeobox protein Meis1 (UniProt: Q60954-1; also known as Myeloid ecotropic viral integration site 1) Homeobox protein Meis2 (UniProt: P97367-2; also known as Meis1-related protein 1) are encoded by the Meis1 and Meis2 (also known as Mrg1, Stra10) genes (Gene ID: 17268 and 17536) in murine species. Meis proteins act as transcription factors that are orthologous to the Drosophila homothorax (Hth) protein. They are expressed dynamically in several organs during embryogenesis and exert potent regulatory activity through their interaction with Hox proteins and other transcription factors. Meis protein contain a TALE (three-amino-acid loop extension) sub-class of the homeodomain that binds to DNA. They directly bind to Pbx proteins and the Meis/Pbx protein complex binds to DNA through respective Meis- and Pbx-consensus binding sites to regulate transcription. The Meis/Pbx complex plays important roles during development of multiple organs. In humans and mice, three homologues Meis1, Meis2 and Meis3 have been identified. Meis1 is expressed at high levels in the lung and at lower levels in the heart and brain. On the other hand, Meis 2 displays spatially restricted expression patterns in the developing nervous system, limbs, face, and in various viscera. In adult, it is mainly expressed in the brain and female genital tract, with a different distribution of the alternative splice forms in these organs. Pancreatic beta cells express both Meis1 and 2. Both Meis1 and 2 are expressed at high levels in all stages of embryonic development of mice (day 7 to 17). Meis3 is expressed in pancreatic islets and is reported to control the expression of expression of PDK1, which promotes survival of pancreatic beta-cells. Deficiency of Meis1 or Meis 2 can lead to embryonic lethality. Meis1-deficient mice die around midgestation due to a general failure of definitive hematopoiesis. (Ref.: Machon, O et al. (2015). BMC Dev. Biol. 515:40; Azcoitia, V et al., (2005). Dev. Biol. 280 (2); 307-320).

Specificity

This rabbit polyclonal antibody detects Meis1/2. It targets an epitope with in 14 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of murine Meis1a, Meis2a, and 13 amino acids from Meis2b isoforms.

Application

Anti-Meis1/2 Antibody, Cat. No. ABE2865, is a highly specific rabbit polyclonal antibody that targets Meis1/2 and has been tested in Chromatin Immunoprecipitation (ChIP), Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Meis1/2 in Immunofluorescence applications (Mercader, N., et. al. (2005). Development. 132(18):4131-42; Marcos, S., et. al. (2015). Development. 142(17):3009-20).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Meis1/2 in Chromatin Immunoprecipitation applications (Penkov, D., et. al. (2013). Cell Rep. 3(4):1321-33; Marcos, S., et. al. (2015). Development. 142(17):3009-20).

Western Blotting Analysis: A representative lot detected Meis1/2 in Western Blotting applications (Mercader, N., et. al. (2005). Development. 132(18):4131-42).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293T cells transfected with pCMVTAG2-Flag-Meis2a.

Western Blotting Analysis: 2 µg/mL of this antibody detected Meis1/2 in HEK293T cells transfected with pCMVTAG2-Flag-Meis2a.

Target description

~51 kDa observed; 43.00, 43.79, and 43.06 kDa for Meis1a, Meis2a, and Meis2b, respectively. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service