Skip to Content
Merck
All Photos(1)

Key Documents

ABC328

Sigma-Aldrich

Anti-CLEC2A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

C-type lectin domain family 2 member A, Keratinocyte-associated C-type lectin, KACL, Proliferation-induced lymphocyte-associated receptor, PILAR, UNQ579, PRO19597

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLEC2A(387836)

General description

C-type lectin domain family 2 member A (CLEC2A) is also called proliferation-induced lymphocyte-associated receptor (PILAR) and Keratinocyte-associated C-type lectin (KACL). CLEC2A is involved in modulating T-cell expansion. It is a 32 kDa single-pass type II membrane protein that belongs to the CTL/CTLD superfamily. CLEC2A is mainly expressed in skin and highly expressed in CD8(+), B lymphocytes and naive CD4(+) T cells. Modulation of CLEC2A signaling may be important for treatment of autoimmune diseases and cancer. CLEC2A is associated with expansion of TCR-stimulated T-cells, modulates the capacity of T-cells to home to lymph nodes through SELL, and facilitates immune recognition of keratinocytes by stimulating natural killer cell mediated cytotoxicity.

Immunogen

KLH-conjugated linear peptide corresponding to human CLEC2A.

Application

Anti-CLEC2A Antibody is a highly specific rabbit polyclonal antibody, that targets CLEC2A & has been tested in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in K562 cell lysate.

Western Blotting/Peptide Inhibition Analysis: 1 µg/mL of this antibody detected CLEC2A in 15 µg of K562 cell lysate.

Target description

~32 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 2 isoforms at ~19 kDa and ~20 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service