Skip to Content
Merck
All Photos(1)

Key Documents

AB5388

Sigma-Aldrich

Anti-Synphilin-1 Antibody, a.a. 829-847

serum, Chemicon®

Synonym(s):

Alpha-Synuclein Interacting Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SNCAIP(9627)

Specificity

Synphilin-1. Immunohistochemical analysis of human brain indicates a high level of specificity. Peptide sequence used shows 88% sequence homology with mouse synphilin-1 but mouse has not been tested for reactivity.

Immunogen

A synthetic peptide from human synphilin-1 (amino acids 829-847).
Epitope: a.a. 829-847

Application

Anti-Synphilin-1 Antibody, a.a. 829-847 detects level of Synphilin-1 & has been published & validated for use in IH.
Immunohistochemistry: 1:1,000-1:2,000.

Note: The antibody has also been used successfully for Western blot but results may vary from lab to lab. The usefulness of the antibody for Western blot must be determined by each individual laboratory.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50 μL of sterile distilled water. Centrifuge if necessary to remove any residue.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C for up to 6 months. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jada H Vaden et al.
Journal of neurochemistry, 148(3), 386-399 (2018-11-20)
Ubiquitin is an essential signaling protein that controls many different cellular processes. While cellular ubiquitin levels normally cycle between pools of free and conjugated ubiquitin, the balance of these ubiquitin pools can be shifted by exposure to a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service