Skip to Content
Merck
All Photos(1)

Key Documents

967107

Supelco

Stainless steel clamp for filling nozzle attachment to walls

Synonym(s):

Clamp for filling nozzle, wall

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£133.00

£133.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
£133.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.03

£133.00


Please contact Customer Service for Availability

Request a Bulk Order

shipped in

ambient

storage temp.

room temp

General description

Safety and convenient handling are the basis for an important part of our very special product developments. For individual solvents installations that end with a flexible filling nozzle inside a fume hood, a clamp to store the filling nozzle safely at the wall is now available.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service