Skip to Content
Merck
All Photos(1)

Documents

8.55072

Sigma-Aldrich

p-Nitrophenyl carbonate Merrifield resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352005
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: amine reactive

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

15-25°C

General description

Resin-bound p-nitrophenyl carbonate esters react readily with amines to provide the corresponding resin-bound carbamate. Leznoff [1] was the first to demonstrate the utility of such resins in solid phase synthesis with the preparation of mono-acylated diamines from symmetrical diamines. Dress man, et al. [2] have used carbamate linked resin-bound amino acid amides to make hydantoins via a base-mediated cyclization/ cleavage strategy; a similar approach has also been used to prepare quinazolidine-2,4-diones [3]. Gordon, et al. [4] have used p-nitrophenyl carbonate Merrifield resin to immobilize a ketone linker through a phenyl carbonate linkage. Loading of this resin can be quantified by measuring the release of p-nitrophenolate [5].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] D. M. Dixit, et al. (1978) Israel J. Chem., 17, 248.
[2] B. A. Dressman, et al. (1996) Tetrahedron Lett., 37, 937.
[3] L. Gouilleux, et al. (1996) Tetrahedron Lett., 37, 7031.
[4] K. Gordon, et al. (2000) Tetrahedron Lett., 41, 8621.
[5] A. Paio, et al. (2003) Tetrahedron Lett.,44, 1867.

Linkage

Replaces: 01-64-0131

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (photometric determination of p-nitrophenyl released upon treatment with piperidin): 0.80 - 1.40 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly ( styrene - 1% DVB ), 100 - 200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service