Skip to Content
Merck
All Photos(1)

Key Documents

8.52346

Sigma-Aldrich

Fmoc-Sec(pMeOBzl)-OH

≥98.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Sec(pMeOBzl)-OH, Fmoc-Sec(Mob)-OH,Fmoc-S-4-methoxybenzyl selenocysteine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
£200.00
250 MG
£381.00
1 G
£1,140.00

£200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
£200.00
250 MG
£381.00
1 G
£1,140.00

About This Item

Empirical Formula (Hill Notation):
C26H25NO5Se
CAS Number:
Molecular Weight:
510.44
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

£200.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Fmoc-Sec(pMeOBzl)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

[Se](C[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O)Cc1ccc(cc1)OC

InChI

1S/C26H25NO5Se/c1-31-18-12-10-17(11-13-18)15-33-16-24(25(28)29)27-26(30)32-14-23-21-8-4-2-6-19(21)20-7-3-5-9-22(20)23/h2-13,23-24H,14-16H2,1H3,(H,27,30)(H,28,29)/t24-/m0/s1

InChI key

XCPAYIZRJRMXBI-DEOSSOPVSA-N

General description

Building block for the introduction of selenocysteine during Fmoc SPPS [1,2,3]. Selenocysteine derivatives can undergo enantiomerization during coupling and can form dehydroalanine and β-piperidinylalanine containing side products during subsequent chain elongation [3]. Therefore, activation methods, such as HBTU or PyBOP which involve the addition of a tertiary, base should be avoided for addition of the Sec and all subsequent residues. Cleavage and side-chain deprotection of Sec-containing peptides can be effected using tFA-m-cresol-thioanisole-EDT-water (80:5:5:5:5)[2] or TFA-water-DCM-TIS (89:5:51) at 4 °C [3]. As the Se-pMeOBzl bond is stable to TFA, these methods result in the formation of the corresponding Sev(pMeOBzl) peptide. Peptides containing two Sec(pMeOBzl) residues can be oxidized directly to the corresponding selenocystinyl peptides by treatment with 5-10% DMSO in TFA [1-3].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] T. Koide, et al. (1993) Chem. Pharm. Bull., 41, 502.
[2] T. Koide, et al. (1993) Chem. Pharm. Bull., 41, 1596.
[3] D. Besse & L. Moroder (1997) J. Peptide Sci., 3, 442.

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service