Skip to Content
Merck
All Photos(1)

Documents

8.45055

Sigma-Aldrich

Pyrrole

Synonym(s):

Pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N
CAS Number:
Molecular Weight:
67.09
MDL number:
UNSPSC Code:
12352005

vapor pressure

8.7 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

550 °C

potency

137 mg/kg LD50, oral (Rat)

expl. lim.

3.10-14.8 % (v/v)

pH

>6 (20 °C, 10 g/L in H2O)

bp

130-131 °C/1013 hPa

mp

-24 °C

transition temp

flash point 39 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Analysis Note

Colour (visual): yellow
Appearance of substance (visual): liquid
Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.967 - 0.971
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service