Skip to Content
Merck
All Photos(1)

Key Documents

8.22189

Sigma-Aldrich

2-Vinylpyridine

(stabilized with 4-tert-butylpyrocatechol) for synthesis

Synonym(s):

2-Vinylpyridine, 2-Ethenyl-pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N
CAS Number:
Molecular Weight:
105.14
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-879-8

vapor pressure

2.5 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

440 °C

potency

100 mg/kg LD50, oral (Rat)

expl. lim.

1.3-10.7 % (v/v)

pH

8 (20 °C, 100 g/L in H2O)

bp

158-160 °C/1013 hPa

mp

-50 °C

transition temp

flash point 51 °C

density

0.97 g/cm3 at 20 °C

storage temp.

−20°C (−15°C to −25°C)

InChI

1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2

InChI key

KGIGUEBEKRSTEW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.974 - 0.976
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

50 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service