Skip to Content
Merck
All Photos(1)

Key Documents

8.18661

Sigma-Aldrich

2-tert-Butylamino-ethanol

for synthesis

Synonym(s):

2-tert-Butylamino-ethanol, N-tert-Butylethanolamin

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
£234.00

£234.00


Please contact Customer Service for Availability


Select a Size

Change View
100 G
£234.00

About This Item

Linear Formula:
(CH3)3CNHCH2CH2OH
CAS Number:
Molecular Weight:
117.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
225-034-5
NACRES:
NA.22

£234.00


Please contact Customer Service for Availability

Quality Level

Assay

≥99.0% (GC)

potency

1620 mg/kg LD50, oral (Rat)
635 mg/kg LD50, skin (Rabbit)

bp

176-177 °C/1013 hPa

mp

40-43 °C

transition temp

flash point 69 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C(C)(C)C)CCO

InChI

1S/C6H15NO/c1-6(2,3)7-4-5-8/h7-8H,4-5H2,1-3H3

InChI key

IUXYVKZUDNLISR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service