Skip to Content
Merck
All Photos(1)

Key Documents

8.18100

Sigma-Aldrich

Pentafluorophenol

for synthesis

Synonym(s):

Pentafluorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6HF5O
CAS Number:
Molecular Weight:
184.06
MDL number:
UNSPSC Code:
12352104
EC Index Number:
212-235-8

Assay

≥99.0% (GC)

form

solid

reaction suitability

reaction type: Addition Reactions

mp

34-36 °C

transition temp

flash point 72 °C

application(s)

peptide synthesis

storage temp.

2-30°C

InChI

1S/C6HF5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

XBNGYFFABRKICK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 32 °C
Melting range (upper value): ≤ 35 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service