Skip to Content
Merck
All Photos(1)

Key Documents

8.07346

Sigma-Aldrich

Platinum(IV) oxide hydrate

80% Pt, hydrogenation catalyst for synthesis

Synonym(s):

Platinum(IV) oxide hydrate (80% Pt), Platinum dioxide, Adam′s catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
£149.00
1 G
£227.00

£149.00


Please contact Customer Service for Availability


Select a Size

Change View
500 MG
£149.00
1 G
£227.00

About This Item

Empirical Formula (Hill Notation):
O2Pt · xH2O
CAS Number:
Molecular Weight:
227.08 (anhydrous basis)
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-223-0
NACRES:
NA.22

£149.00


Please contact Customer Service for Availability

Product Name

Platinum(IV) oxide hydrate (80% Pt), hydrogenation catalyst for synthesis

Quality Level

mp

>450 °C

storage temp.

2-30°C

SMILES string

[Pt](=O)=O.O

InChI

1S/H2O.2O.Pt/h1H2;;;

InChI key

SPGAMGILENUIOF-UHFFFAOYSA-N

Analysis Note

Assay (of Pt): 80.0 - 84.0 %
Suitable for hydrogenation catalyst: passes test
Identity ( RFA): passes test

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service