Skip to Content
Merck
All Photos(1)

Key Documents

8.07167

Sigma-Aldrich

Nonanoic acid

for synthesis

Synonym(s):

Nonanoic acid, Pelargonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
£31.20
1 L
£50.50

£31.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
£31.20
1 L
£50.50

About This Item

Linear Formula:
CH3(CH2)7COOH
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-931-2
NACRES:
NA.22

£31.20


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.04 hPa ( 20 °C)

Quality Level

Assay

≥90.0% (GC)

form

liquid

autoignition temp.

405 °C

expl. lim.

0.8-9 % (v/v)

pH

4.4 (25 °C, 0.1 g/L in H2O)

kinematic viscosity

9.0 cSt(20 °C)

mp

13 °C

transition temp

flash point 137 °C

solubility

0.3 g/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCCCCCC

InChI

1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)

InChI key

FBUKVWPVBMHYJY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.904 - 0.907
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service