Skip to Content
Merck
All Photos(1)

Documents

8.06807

Sigma-Aldrich

4-Nitrotoluene

for synthesis

Synonym(s):

4-Nitrotoluene, p-Nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(NO2)C6H4CH3
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352102
EC Index Number:
202-808-0

vapor pressure

0.4 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

solid

autoignition temp.

456 °C

potency

1960 mg/kg LD50, oral (Rat)
>16000 mg/kg LD50, skin (Rat)

expl. lim.

1.6 % (v/v)

bp

238-239 °C/1013 hPa

mp

50-52 °C

transition temp

flash point 106 °C

solubility

0.26 g/L

density

1.39 g/cm3

bulk density

700 kg/m3

storage temp.

2-30°C

InChI

1S/C7H7NO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3

InChI key

ZPTVNYMJQHSSEA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 50 °C
Melting range (upper value): ≤ 52 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service