Skip to Content
Merck
All Photos(2)

Key Documents

8.01678

Sigma-Aldrich

Bis(2-chloroethyl) ether

for synthesis

Synonym(s):

Bis(2-chloroethyl) ether, 2,2′-Dichlorodiethyl ether, ß,ß′-Dichloroethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
£32.20
1 L
£102.00

£32.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
£32.20
1 L
£102.00

About This Item

Linear Formula:
(ClCH2CH2)2O
CAS Number:
Molecular Weight:
143.01
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-870-1
NACRES:
NA.22

£32.20


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.07 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

365 °C

expl. lim.

0.8 % (v/v)

bp

177-178 °C/1013 hPa

mp

-51.9 °C

transition temp

flash point 55 °C

density

1.22 g/cm3 at 20 °C

SMILES string

ClCCOCCCl

InChI

1S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2

InChI key

ZNSMNVMLTJELDZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.218 - 1.219
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service