Skip to Content
Merck
All Photos(1)

Key Documents

588X

Sigma-Aldrich

Eosin Y, 1% Alcoholic Solution

Harleco®, suitable for histology

Synonym(s):

Counterstain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco®

mp

-97.77 °C

density

0.79 g/cm3

λmax

520-530 nm

application(s)

diagnostic assay manufacturing
hematology

histology

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® cytoplasmic counterstain for H&E in an alcoholic base. 1% PDC

Application

HARLECO® cytoplasmic counterstain for H&E in an alcoholic base. 1% PDC
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Cytoplasmic counterstain for hematoxylin in an alcoholic base. Most widely used in histology for H&E staining of tissues.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service