Skip to Content
Merck
All Photos(1)

Key Documents

559395

Sigma-Aldrich

SB 203580, Hydrochloride

SB 203580, Hydrochloride, CAS 869185-85-3, is a water-soluble form of SB 203580. A highly potent, selective, ATP-competitive inhibitor of p38 MAP Kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).

Synonym(s):

SB 203580, Hydrochloride, 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)1H-imidazole, HCl, p38 MAP Kinase Inhibitor XVI, Hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
£162.00
10 MG
£391.00

£162.00


Estimated to ship on06 May 2025Details



Select a Size

Change View
1 MG
£162.00
10 MG
£391.00

About This Item

Empirical Formula (Hill Notation):
C21H16FN3OS · xHCl
CAS Number:
Molecular Weight:
377.43 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

£162.00


Estimated to ship on06 May 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

bright yellow

solubility

water: 10 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)c2[nH]c(nc2c4ccncc4)c3ccc(cc3)[S](=O)C.Cl

InChI

1S/C21H16FN3OS.ClH/c1-27(26)18-8-4-16(5-9-18)21-24-19(14-2-6-17(22)7-3-14)20(25-21)15-10-12-23-13-11-15;/h2-13H,1H3,(H,24,25);1H

InChI key

WOSGGXINSLMASH-UHFFFAOYSA-N

General description

A water-soluble form of the potent MAP kinase inhibitor SB 203580 (Cat. No. 559389).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p38 MAP kinase
Product does not compete with ATP.
Reversible: no
Target IC50: 34 nM against p38 MAP kinase in vitro, 600 nM in cells; 50-100 nM against IL-1 and TNF-a production from LPS-stimulated human monocytes and the human monocyte cell line THP-1; 0.2-1.0 µM against platelet aggregation caused by collagen

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Merete Lund et al.
Biology of reproduction, 108(2), 258-268 (2022-10-11)
Angiogenesis within the ovarian follicle is an important component of ovulation. New capillary growth is initiated by the ovulatory surge of luteinizing hormone (LH), and angiogenesis is well underway at the time of follicle rupture. LH-stimulated follicular production of vascular
Silvia Fischer et al.
Journal of neuroinflammation, 18(1), 252-252 (2021-11-04)
Astrocytes participate in the local innate immune response of the central nervous system. In response to stress such as ischemia, activated cells release endogenous factors known as damage-associated molecular patterns (DAMPs). Self-extracellular RNA (eRNA) is such a ubiquitous alarm signal.
Alain P Gobert et al.
Cell reports, 33(11), 108510-108510 (2020-12-17)
Innate responses of myeloid cells defend against pathogenic bacteria via inducible effectors. Deoxyhypusine synthase (DHPS) catalyzes the transfer of the N-moiety of spermidine to the lysine-50 residue of eukaryotic translation initiation factor 5A (EIF5A) to form the amino acid hypusine.
Zhaojuan Yang et al.
Nature communications, 12(1), 4852-4852 (2021-08-13)
Oncogenic activation of KRAS and its surrogates is essential for tumour cell proliferation and survival, as well as for the development of protumourigenic microenvironments. Here, we show that the deubiquitinase USP12 is commonly downregulated in the KrasG12D-driven mouse lung tumour
Alice Prigent et al.
Journal of neurochemistry, 148(6), 746-760 (2018-12-28)
The protein alpha-synuclein whose expression is strongly implicated in Parkinson's disease (PD) is not only expressed in the CNS but also in the enteric nervous system (ENS). The growing body of evidence suggesting that gastrointestinal inflammation is involved in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service