Skip to Content
Merck
All Photos(1)

Documents

539481-M

Sigma-Aldrich

PKA, Catalytic Subunit, Mouse, Recombinant, E. coli

Synonym(s):

cAMP-Dependent Protein Kinase, Catalytic Subunit, Protein Kinase A, Catalytic Subunit

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202

form

liquid

Quality Level

specific activity

≥2.5 x 106 U/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

Phosphatases, proteases, none detected

storage temp.

−20°C

General description

Recombinant, mouse PKA catalytic subunit expressed in E. coli. Regulates a number of cellular functions including gene expression, carbohydrate metabolism, Ca2+ homeostasis, and ion channel function. Suitable for studying the phosphorylation of purified substrates and for use with cell extracts. Unlike the holoenzyme, the catalytic subunit does not require cAMP for activation.
Recombinant, mouse PKA catalytic subunit expressed in E. coli. The catalytic subunit of protein kinase A that combines with the regulatory subunit, in the absence of cAMP, to form the inactive PKA holoenzyme. Unlike the holoenzyme, the catalytic subunit does not require cAMP for activation. PKA-Cα is reported to potentiate IP3-induced Ca2+ mobilization in isolated hepatocytes. Suitable for studies of PKA phosphorylation in vitro with purified substrates or following the introduction into intact cells or cell extracts.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will transfer 1.0 pmol phosphate to Kemptide (LRRASLG; Cat. No. 05-23-4900) per min at 30°C, pH 7.5.

Physical form

In 50 mM NaCl, 20 mM Tris-HCl, 1 mM EDTA, 2 mM DTT, 50% glycerol, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Grant, B.D., and Adams, J.A. 1996. Biochemistry35, 2022.
Lee, S-L. and Steinberg, R.A. 1996. J. Biol. Chem.271, 16553.
Adams, J.A., et al. 1995. Biochemistry 34, 2447.
Bosma, M.M., et al. 1993. J. Neurosci.13, 5242.
Brindle, P., et al. 1993. Nature364, 821.
Hajnoczky, G., et al. 1993. Biochem. J.293, 413.
Sculptoreanu, A., et al. 1993. Proc. Natl. Acad. Sci. USA90, 10135.
Wang, L-Y., et al. 1993. Science259, 1173.
Slice, L.W., and Taylor, S.S. 1989. J. Biol. Chem.264, 20940.
Uhler, M.D., et al. 1986. Proc. Natl. Acad. Sci. USA83, 1300.
Walsh, D.A., et al. 1968. J. Biol. Chem.243, 3763.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service