Skip to Content
Merck
All Photos(1)

Key Documents

5.33088

Sigma-Aldrich

Monoacylglycerol Lipase Inhibitor, Cpd21

Synonym(s):

Monoacylglycerol Lipase Inhibitor, Cpd21, MAGL Inhibitor, Cpd21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26O4
CAS Number:
Molecular Weight:
402.48
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

400 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable hexanoate ester compound that acts as a potent, selective, non-competitive, and reversible inhibitor of monoacylglycerol lipase (MAGL, Ki = 400 nM, IC50 = 240 nM for hydrolysis of 2- arachidonoylglycerol (2-AG). Displays good selectivity over a broad panel of related enzymes and does not bind to CB1 and CB2 cannabinoid receptors (Ki >10 µM). Shown to inhibit mouse brain MAGL (IC50 = 180 nM), but does not affect the activity of fatty acid amide hydrolaase (FAAH; IC50 = 59 µM) in any significant manner. Improves the course of experimental autoimmune encephalitis (EAE) murine model of multiple sclerosis (5 mg/kg/day, i.p. for 21 days) without inducing any undesirable CB1-agonist-like activity.
MAGL inhibitor, Monoacylglycerol Lipase Inhibitor

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MAGL

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hernandez-Torres, G., et al. 2014. Angew Chem Int Ed Engl.53, in press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service