Skip to Content
Merck
All Photos(1)

Key Documents

5.30306

Sigma-Aldrich

PPM1D Phosphatase Inhibitor II, GSK2830371

Synonym(s):

PPM1D Phosphatase Inhibitor II, GSK2830371, ( S)-5-((5-Chloro-2-methylpyridin-3-ylamino)methyl)-N-(3-cyclopentyl-1-(cyclopropylamino)-1-oxopropan-2-yl)thiophene-2-carboxamide, PP2Cδ Inhibitor II, Wip1 Inhibitor II, WT-53-Induced Phosphatase Inhibitor; GSK 2830371, (S)-5-((5-Chloro-2-methylpyridin-3-ylamino)methyl)-N-(3-cyclopentyl-1-(cyclopropylamino)-1-oxopropan-2-yl)thiophene-2-carboxamide, PP2Cδ Inhibitor II, Wip1 Inhibitor II, WT-53-Induced Phosphatase Inhibitor; GSK 2830371

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29ClN4O2S
CAS Number:
Molecular Weight:
461.02
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

light yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyridinylaminomethylthienylcarboxamide that acts as a potent inhibitor against wild-type p53-induced phosphatase Wip1/PPM1D/PP2Cδ activity (IC50/[wip1]/[substrate]= 6 nM/1 nM Wip1/50 µM FDP and 13 nM/15 nM Wip1/8 nM pT180 p38) via an allosteric interaction with the ′flap′ region outside the catalytic site in a reversible and substrate-noncompetitive manner, while exhibiting little potency toward yeast YopH and 22 other human phosphatases (IC50 >30 µM), including PPM1A/PP2Cα & PPM1K/PP2Cκ. Shown to increase cellular Wip1 substrates phosphorylation, including p53 (S15), Chk2 (T68), H2AX (S139), and ATM (S1981), leading to p53 response protein p21/Waf1 upregulation and selective growth inhibition against breast cancer MCF-7 (IC50 = 500 nM in 14-d colony formation assay) and 22 lymphoid cell lines (IC50 from 24 nM to 2.2 µM in 7-d viability assay) expressing wt p53, but not BT474 breast cancer or a panel of 25 hematologic cancer cultures with mutant p53 status (IC50 >10 µM). Albeit short in vivo half-life, frequent and high daily oral dosages effectively suppress B-cell lymphoma DoHH2-derived tumor growth in mice (by 41% and 68% in 14 days with 75 mg/kg/12 h or 150 mg/kg/8 h oral dosage, respectively) in vivo. In addition to direct inhibition against Wip1 phosphatase activity, GSK2830371 binding also results in downregulation of cellular Wip1 protein, but not mRNA, level due to ubiquitination-dependent degradation. MG-132 (Cat. Nos. 474790, 474787, 474788, and 474791) effectively prevents GSK2830371-induced Wip1 degradation without interfering with the phosphorylation enhancing effect of GSK2830371 toward Wip1 substrates.
A cell-permeable, orally available pyridinylaminomethylthienylcarboxamide compound that acts as a potent, allosteric, non-competitive, and reversible inhibitor of wild-type p53-induced phosphatase (Wip1; IC50 = 6 nM for dephosphorylation of fluorescein diphosphate by Wip1 (2-240) and 13 nM for phospho-p38 MAP kinase (Thr180)). Does not affect the activity of 21 other phosphatases studied (IC50 ≥ 30 µM). Binds to the flap region in close proximity to Wip1 catalytic site and impairs ubiquitin-mediated degradation of Wip1. Co-treatment of cells with MG-132 (Cat. No. 474790) reverses the reduction in Wip1 levels in cells. Enhances phosphorylation of Chk2 (Thr68), p53 (Ser15), H2AX (Ser139), and ATM (Ser1981) and upregulates the levels of p21/WAF1 in various tumor cells. Shown to block the growth of hematopoietic tumor cell lines and Wip1-amplified breast tumor cells harboring wild-type TP53, but does not affect BT474 cells carrying mutant p53 and amplified Wip1. Due to its short half life and reversible action, a sustained inhibition is warranted to block tumor growth. Reduces the growth of DOHH2 lymphoma tumor xenografts in mice (~150 mg/kg, p.o. bid or tid).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Wip1/PPM1D/PP2Cδ
Reversible: yes
Target IC50: 6 nM for dephosphorylation of fluorescein diphosphate by Wip1 (2-240) and 13 nM for phospho-p38 MAP kinase (Thr180)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gilmartin, A.G., et al. 2014. Nature Chem. Biol.10, 181.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service