Skip to Content
Merck
All Photos(1)

Key Documents

529627

Sigma-Aldrich

PPIase-Parvulin Inhibitor - CAS 64005-90-9 - Calbiochem

The PPIase-Parvulin Inhibitor, also referenced under CAS 64005-90-9, controls the biological activity of PPIase-Parvulin.

Synonym(s):

PPIase-Parvulin Inhibitor - CAS 64005-90-9 - Calbiochem, PiB, Diethyl-1,3,6,8-tetrahydro-1,3,6,8-tetraoxobenzo[lmn][3,8]phenanthroline-2,7-diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N2O8
CAS Number:
Molecular Weight:
438.39
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brownish

solubility

chloroform: 1 mg/mL
ethanol: 250 μg/mL, cloudy (suspension)

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H18N2O8/c1-3-31-15(25)9-23-19(27)11-5-7-13-18-14(8-6-12(17(11)18)20(23)28)22(30)24(21(13)29)10-16(26)32-4-2/h5-8H,3-4,9-10H2,1-2H3

InChI key

WNKQGFNIIHNGQM-UHFFFAOYSA-N

General description

A cell-permeable napthalenetetracarboxylic-bisimide compound that displays anti-proliferative properties (IC50<5 µM against a panel of human cancer cell lines). Causes cell cycle arrest and acts as a potent, selective, reversible, competitive, active site binding inhibitor of Pin1/Par14 PPIase (peptidyl prolyl cis-trans isomerase) activity (IC50 = ~ 1.5 µM). Does not bind to DNA or functions as a topoisomerase I inhibitor.
A cell-permeable, napthalenetetracarboxylic-bismide compound that displays anti-proliferative properties (IC50<5 µM against a panel of human cancer cell lines). Causes cell cycle arrest and acts as a potent, selective, reversible, competitive, active site-binding inhibitor of Pin1/Par14 peptidylprolyl cis-trans isomerase (PPIase) activity (IC50 ~ 1.5 µM). Does not bind to DNA or function as a topoisomerase I inhibitor.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Pin1/Par14 PPIase (peptidyl prolyl cis-trans isomerase) activity
Product competes with ATP.
Reversible: yes
Target IC50: <5 µM for anti-proliferative properties against a panel of human cancer cell lines; ~ 1.5 µM inhibiting Pin1/Par14 PPIase (peptidyl prolyl cis-trans isomerase) activity

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Uchida, T., et al. 2003. Chem. Biol.10, 15.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Napoletano et al.
Cell reports, 36(11), 109694-109694 (2021-09-16)
Chromatin organization plays a crucial role in tissue homeostasis. Heterochromatin relaxation and consequent unscheduled mobilization of transposable elements (TEs) are emerging as key contributors of aging and aging-related pathologies, including Alzheimer's disease (AD) and cancer. However, the mechanisms governing heterochromatin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service