Skip to Content
Merck
All Photos(1)

Key Documents

513024-M

Millipore

PD 151746

A cell-permeable, non-peptidic, and highly selective calpain inhibitor directed towards the calcium binding sites of calpain.

Synonym(s):

PD 151746, 3-(5-Fluoro-3-indolyl)-2-mercapto-(Z)-2-propenoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8FNO2
CAS Number:
Molecular Weight:
205.19
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 200 mg/mL
methanol: 5 mg/mL

storage temp.

−20°C

InChI

1S/C11H8FNO2S/c12-7-1-2-9-8(4-7)6(5-13-9)3-10(16)11(14)15/h1-5,13,16H,(H,14,15)/b10-3-

InChI key

HWMQHECFXSVZGN-KMKOMSMNSA-N

General description

A cell-permeable, non-peptidic, and highly selective calpain inhibitor directed towards the calcium binding sites of calpain. Displays over 20-fold greater selectivity for calpain-1 (Cat. Nos. 208712 and 208713) over calpain-2 (Cat. Nos. 208715 and 208718) (Ki values: 260 nM and 5.33 µM, respectively). Has been shown to prevent cycloheximide-induced apoptosis.

Biochem/physiol Actions

Primary Target
calpain-1
Target Ki: 260 nM against calpain-1

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Squier, M.K., et al. 1999. J. Cell Physiol.178, 311.
Wang, K.K., et al. 1996. Proc. Natl. Acad. Sci. USA93, 6687.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service