Skip to Content
Merck
All Photos(1)

Key Documents

5.00556

Sigma-Aldrich

Tankyrase1/2 Inhibitor V, WIKI4

Synonym(s):

Tankyrase1/2 Inhibitor V, WIKI4, 2-(3-((4-(4-methoxyphenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)sulfanyl)propyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione, Wnt Pathway Inhibitor XVIII, TNKS1/2 Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H23N5O3S
CAS Number:
Molecular Weight:
521.59
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

15 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

SMILES string

COC1=CC=C(C=C1)N2C(=NN=C2SCCCN3C(=O)C4=CC=CC5=C4C(=CC=C5)C3=O)C6=CC=NC=C6

General description

A cell-permeable triazole derivative that acts as a reversible TNKS inhibitor (IC50 = 15 nM in cell-free TNKS2 auto-ribosylation assays) and selectively inhibits Wnt/β-catenin, but not NF-κB, TGFβ, or retinoic acid, signaling pathway-dependent reporter activity. Although structurally distinct from XAV939 (Cat. No. 575545), both compounds are shown to induce basal axin1/2 protein accumulation in DLD1 colon cancer cells with mutant APC (2.5 µM; 2 to 4 h) and prevent Wnt3a-induced axin1 protein degradation and β-catenin accumulation in A375 melanoma cells (1 µM; and up to 24 h) with similar potency. Likewise, WIKI4 also effectively inhibits the Wnt/β-catenin pathway-dependent growth of DLD1 (37% and 67% inhibition of colony formation in 10 days, respectively, in the presence of 0.1 and 1 µM WIKI4).
A cell-permeable triazole derivative that acts as a reversible TNKS inhibitor (IC50 = 15 nM) and selectively inhibits Wnt/β-catenin, but not NF-κB, TGFβ, or retinoic acid, signaling pathway-dependent transcription activity. Both WIKI4 and XAV939 (Cat. No. 575545) are shown to induce basal axin1/2 protein accumulation in DLD1 colon cancer cells (2.5 µM; 2 to 4 h) and prevent Wnt3a-induced axin1 protein degradation and β-catenin accumulation in A375 melanoma cells (1 µM; up to 24 h) with similar potency. Likewise, WIKI4 also effectively inhibits the Wnt/β-catenin pathway-dependent growth of DLD1 (by 67% in 10 days;1 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TNKS2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Followig reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

James, R.G., et al. 2012. PLoS. One.7, e50457.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service