Skip to Content
Merck
All Photos(1)

Key Documents

420413

Millipore

Polymyxin B Sulfate, Sterile-Filtered Aqueous Solution, Cell Culture Tested

An antibiotic that inhibits phospholipid sensitive Ca2+-dependent protein kinase.

Synonym(s):

Polymyxin B Sulfate, Sterile-Filtered Aqueous Solution, Cell Culture Tested, Aerosporin, Sterile-Filtered Aqueous Solution, Cell Culture Tested

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

General description

An antibiotic that inhibits phospholipid sensitive Ca2+-dependent protein kinase. Mixture of polymyxin B1 sulfate and polymyxin B2 sulfate. Binds to cell wall and makes it more permeable, causing fluid uptake. Effective against Gram-negative bacteria.

Biochem/physiol Actions

≥7500 units/mg

Warning

Toxicity: Harmful (C)

Physical form

Supplied at 50 mg/ml in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Lucas, M., et al. 1994. Biochem. Pharmacol.47, 667.
Ngezahayo, A. and Kolb, H.A. 1993. Pflugers Arch.422, 413.
Kubo, M. and Okada, Y. 1992. J. Physiol.456, 351.
Strasser, R.H., et al. 1992. Circ. Res.70, 1304.
Hegemann, L., et al. 1991. Eur. J. Pharmacol.207, 17.
Inaba, H. and Finkins, J.P. 1991. Am. J. Physiol.261, R26.
Marra, M.N., et al. 1990. J. Immunol.144, 662.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service