Skip to Content
Merck
All Photos(1)

Key Documents

407248

Sigma-Aldrich

IGF-1R Inhibitor II, PQ401

The IGF-1R Inhibitor II, also referenced under CAS 196868-63-0, controls the biological activity of IGF-1R. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

IGF-1R Inhibitor II, PQ401, N-(2-Methoxy-5-chlorophenyl)-Nʹ-(2-methylquinolin-4-yl)-urea, PQ401

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16ClN3O2
CAS Number:
Molecular Weight:
341.79
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H16ClN3O2/c1-11-9-15(13-5-3-4-6-14(13)20-11)21-18(23)22-16-10-12(19)7-8-17(16)24-2/h3-10H,1-2H3,(H2,20,21,22,23)

InChI key

YBLWOZUPHDKFOT-UHFFFAOYSA-N

General description

A cell-permeable phenylquinolinyl-urea compound that inhibits IGF-1R autophosphorylation (IC50 = 12 µM in inhibiting ligand-induced autophosphorylation in MCF-7 cells and <1 µM in a cell-free kinase assay). Reduces IGF-1R-dependent tumor cell growth both in vitro (IC50 = 8 and 15 µM for MCF-7 and MCNeuA, respectively) and in mice in vivo (80% reduction, 100 mg/kg thrice a week per i.p.). The mechanism of inhibition probably involves indirect blocking of ATP binding.
A cell-permeable phenylquinolinyl-urea compound that inhibits IGF-1R autophosphorylation (IC50 = 12 µM in inhibiting ligand-induced autophosphorylation in MCF-7 cells and <1 µM in a cell-free kinase assay). Reduces IGF-1R-dependent tumor cell growth both in vitro (IC50 = 8 and 15 µM for MCF-7 and MCNeuA, respectively) and in mice in vivo (80% reduction, 100 mg/kg thrice a week, i.p.). The mechanism of inhibition probably involves indirect blocking of ATP binding.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IGF-1R
Product does not compete with ATP.
Reversible: no
Target IC50: 12 µM in inhibiting ligand-induced IGF-1R autophosphorylation in MCF-7 cells and <1 µM in a cell-free kinase assay; 8 and 15 µM in reducing IGF-1R-dependent tumor cell growth both in vitro for MCF-7 and MCNeuA, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gable, K.L., et al. 2006. Mol. Cancer Ther.5, 1079.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service