Skip to Content
Merck
All Photos(1)

Key Documents

376

Sigma-Aldrich

Wright Stain Powder

Harleco®, certified by the BSC

Synonym(s):

Wright Stain Powder, Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116121
EC Index Number:
273-541-5

Agency

certified by the BSC

Quality Level

form

solid

manufacturer/tradename

Harleco®

λmax

520-530 nm (Eosin)
645-655 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C20H8Br4O5.C16H18N3S/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13/h1-6H,(H2-,25,26,27,28);5-10H,1-4H3/q;+1/p+1

InChI key

AXIKDPDWFVPGOD-UHFFFAOYSA-O

General description

HARLECO® For differential blood staining
HARLECO® Wright Stain
376-12 25g Glass Bottle

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used extensively in staining blood cells.
HARLECO® For differential blood staining

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service