Skip to Content
Merck
All Photos(1)

Key Documents

375228

Sigma-Aldrich

Hepatocyte Growth Factor, Human, Recombinant, S. frugiperda

Synonym(s):

rhHGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg endotoxin (EU/μg HGF)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human hepatocyte growth factor expressed in Spodoptera frugiperda insect cells. Pleiotropic growth factor that has also been called scatter factor, hepatopoietin A, and mammary growth factor. Through binding to its high affinity met receptor, HGF has marked and varied effects on hepatocytes and other epithelial and endothelial cell types. HGF-induced proliferation of primary hepatocytes is mediated by activation of PI 3-kinase. Acts as a potent mitogen for mature hepatocytes.

Biochem/physiol Actions

ED₅₀ = 20-40 ng/ml as measured by its ability to stimulate ³H-thymidine incorporation in an HGF-responsive epithelial cell line, 4MBr-5

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a sterile-filtered solution in 20 mM Sodium Phosphate, 0.5 M NaCl, 50 µg BSA/µg HGF, pH 7.0.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long term storage or refrigerate (4°C) for short term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for 1 month at 4°C or for up to 3 months at -70°C.
Reconstitute to a concentration of ≥1 µg/ml in sterile PBS containing ≥0.1% HSA or BSA.

Other Notes

Kaido, T., et al. 1996. Biochem. Biophys. Res. Commun. 218, 1.
Kobayashi, Y., et al. 1996. Biochem. Biophys. Res. Commun. 220, 7.
Skouteris, G.G., et al. 1996. Biochem. Biophys. Res. Commun. 218, 229.
Rubin, J.S., et al. 1993. Biochim. Biophys. Acta 1155, 357.
Strain, A.J. 1993. J. Endocrinol. 137, 1. Nakamura, T., et al. 1989. Nature 342, 440.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maren Schulze et al.
Methods in molecular biology (Clifton, N.J.), 329, 45-58 (2006-07-19)
The in vitro differentiation of mouse embryonic stem (ES) cells into different somatic cell types such as neurons, endothelial cells, or myocytes is well established, and many mouse ES cell lines have been created so far. The establishment of rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service