361558
GSK-3 Inhibitor XV - Calbiochem
The GSK-3 Inhibitor XV controls the biological activity of GSK-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
GSK-3 Inhibitor XV - Calbiochem
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
purple
solubility
DMSO: 6 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
The racemic mixture of a cell-permeable pyridocarbazolo-cyclopentadienyl Ruthenium complex that acts as a reversible, ATP-competitive, and highly potent inhibitor of GSK-3 (IC50 = 0.4 and 0.6 nM for GSK-3α and GSK-3β, respectively) with ∼5-fold selectivity over PIM-1 kinase (IC50 = 3 nM). Studies using isolated enantiomers have shown that the (R)-enantiomer is GSK-3-selective (IC50 = 0.35, 0.55, and 35 nM for GSK-3α GSK-3β, and Pim-1, respectively) and interferes with normal zebrafish embryo development (1 µM), while the (S)-enantiomer is Pim-1-selective (IC50 = 80, 90, and 3 nM for GSK-3α GSK-3β, and Pim-1, respectively) and exhibits no effect in embryo development at the same dosage.
Biochem/physiol Actions
Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 0.4 and 0.6 nM for GSK-3α and GSK-3β, respectively
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Atilla-Gokcumen, G.E., et al. 2006. ChemBioChem7, 1443.
Legal Information
Sold for research purposes only, pursuant to an agreement with University of Pennsylvania.Sold under license of U.S. Patent 7,488,817.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Neurobiology of disease, 136, 104720-104720 (2019-12-28)
Parkinson's disease (PD) is a currently incurable disease and the number of patients is expected to increase due to the extended human lifespan. α-Synuclein is a pathological hallmark of PD and variations and triplications of the gene encoding α-synuclein are
Chembiochem : a European journal of chemical biology, 7(9), 1443-1450 (2006-07-22)
A chiral second-generation organoruthenium half-sandwich compound is disclosed that shows a remarkable selectivity and cellular potency for the inhibition of glycogen synthase kinase 3 (GSK-3). The selectivity was evaluated against a panel of 57 protein kinases, in which no other
Molecular biology and evolution, 37(2), 507-523 (2019-10-22)
Evolution of highly invasive placentation in the stem lineage of eutherians and subsequent extension of pregnancy set eutherians apart from other mammals, that is, marsupials with short-lived placentas, and oviparous monotremes. Recent studies suggest that eutherian implantation evolved from marsupial
Brain pathology (Zurich, Switzerland), 31(1), 103-119 (2020-07-28)
Deposition of extensively hyperphosphorylated tau in specific brain cells is a clear pathological hallmark in Alzheimer's disease and a number of other neurodegenerative disorders, collectively termed the tauopathies. Furthermore, hyperphosphorylation of tau prevents it from fulfilling its physiological role as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service