360589-M
Tetrahydrofuran
≥99.0%, ACS reagent, contains 250 ppm BHT as inhibitor
Synonym(s):
THF, Butylene oxide, Oxolane, Tetramethylene oxide
About This Item
143 mmHg ( 20 °C)
Recommended Products
Product Name
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
grade
ACS reagent
vapor density
2.5 (vs air)
vapor pressure
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
Assay
≥99.0%
autoignition temp.
610 °F
contains
250 ppm BHT as inhibitor
expl. lim.
1.8-11.8 %
impurities
≤0.015% peroxide (as H2O2)
≤0.05% water
evapn. residue
≤0.03%
color
APHA: ≤20
refractive index
n20/D 1.407 (lit.)
bp
65-67 °C (lit.)
mp
−108 °C (lit.)
solubility
water: soluble
density
0.889 g/mL at 25 °C (lit.)
SMILES string
C1CCOC1
InChI
1S/C4H8O/c1-2-4-5-3-1/h1-4H2
InChI key
WYURNTSHIVDZCO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- RAFT (Reversible Addition-Fragmentation chain Transfer) polymerization of p-acetoxystyrene.[2]
- Synthesis of di-tert-butyl-phosphinoferrocene.[3]
- Synthesis of n-TiO2-based amphiphilic polymer brushes.[4]
- To dissolve the LiCl from the Mg2Si nanoparticles.[5]
Other Notes
This product will be phased out towards early 2019. There is a replacement product available : 360589
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.00 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service