Skip to Content
Merck
All Photos(1)

Documents

36-017

Sigma-Aldrich

Anti-cleaved-Tau (Asp421) Antibody, clone C3

clone C3, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C3, monoclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

Specificity

cleaved-Tau

Immunogen

Synthetic peptide corresponding to amino acids 412-421(CSSTGSIDMVD) of human Tau with a Cys at the N-terminal end

Application

Anti-cleaved-Tau (Asp421) Antibody, clone C3 is an antibody against cleaved-Tau (Asp421) for use in IP, WB, IH.

Quality

routinely evaluated by immunoblot on Tau fusion protein cleaved by Caspase 3 (Catalog #14-264)

Target description

50-70kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol
Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Biundo et al.
Translational psychiatry, 7(8), e1198-e1198 (2017-08-09)
TAU mutations are genetically linked to fronto-temporal dementia (FTD) and hyper-phosphorylated aggregates of Tau form neurofibrillary tangles (NFTs) that constitute a pathological hallmark of Alzheimer disease (AD) and FTD. These observations indicate that Tau has a pivotal role in the
Urmi Sengupta et al.
Methods in molecular biology (Clifton, N.J.), 1779, 113-146 (2018-06-11)
An increasing number of studies have demonstrated the existence of multiple conformational entities of tau, as have been observed for prion protein. We have developed and optimized techniques to isolate and study oligomeric tau strains both in vitro and ex
Modeling tau polymerization in vitro: a review and synthesis.
Gamblin, T Chris, et al.
Biochemistry, 42, 15009-15017 (2003)
Franck R Petry et al.
Methods in molecular biology (Clifton, N.J.), 1523, 263-272 (2016-12-16)
In Alzheimer's disease and other tauopathies, tau displays several abnormal post-translation modifications such as hyperphosphorylation, truncation, conformation, and oligomerization. Mouse monoclonal antibodies have been raised against such tau modifications for research, diagnostic, and therapeutic purposes. However, many of these primary
Michelle A Utton et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(15), 6394-6400 (2002-08-02)
We demonstrate that the microtubule-associated protein tau, in the form of enhanced green fluorescent protein (EGFP) tau, is transported along axons of neurons in culture in the slow component of axonal transport with a speed comparable with that previously measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service