Skip to Content
Merck
All Photos(1)

Key Documents

347436

Sigma-Aldrich

GGACK

An irreversible inhibitor of urokinase (IC₅₀ 1 µM) and Factor Xa.

Synonym(s):

GGACK, Glu-Gly-Arg-chloromethyl Ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£196.00

£196.00


Estimated to ship on06 May 2025Details



Select a Size

Change View
5 MG
£196.00

About This Item

Empirical Formula (Hill Notation):
C14H25ClN6O5
Molecular Weight:
392.84
UNSPSC Code:
12352200
NACRES:
NA.77

£196.00


Estimated to ship on06 May 2025Details


Quality Level

Assay

≥80% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

An irreversible inhibitor of urokinase (IC50<1 µM) and Factor Xa.

Biochem/physiol Actions

Cell permeable: no
Primary Target
urokinase
Product does not compete with ATP.
Reversible: no
Target IC50: <1 µM against urokinase

Warning

Toxicity: Standard Handling (A)

Sequence

H-Glu-Gly-Arg-CMK

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C. Avoid freeze/thaw cycles of stock solution.

Other Notes

Kettner, C., and Shaw, E. 1979. Biochim. Biophys. Acta569, 31.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service