Skip to Content
Merck
All Photos(1)

Documents

344079-M

Millipore

Flurbiprofen

A mixture of S(+) and R(-) enantiomers.

Synonym(s):

Flurbiprofen, (±)-2-Fluoro-α-methyl[1,1ʹ-biphenyl]-4-acetic Acid, U-27182

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13FO2
CAS Number:
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL

storage temp.

10-30°C

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)

InChI key

SYTBZMRGLBWNTM-UHFFFAOYSA-N

General description

A mixture of S(+) and R(–) enantiomers. A cell-permeable, non-steroidal anti-inflammatory agent that acts as a potent inhibitor of cyclooxygenase (IC50 = 5 nM for LPS-induced COX in human peripheral blood cells). Strongly inhibits insoluble transthyretin (TTR) amyloid fibril formation. Suppresses iNOS expression in RAW 264.7 macrophages. Reduces microglial activation and β-amyloid deposit in APP+PS1 transgenic mice.
A mixture of S(+) and R(-) enantiomers. A cell-permeable, non-steroidal anti-inflammatory (NSAID) agent that acts as a potent inhibitor of cyclooxygenase (IC50 = 5 nM for LPS-induced COX in human peripheral blood cells). The S(+) enantiomer is reported to be more potent as a COX-2 inhibitor than the R(-) enantiomer. Suppresses iNOS expression in RAW 264.7 macrophages. Reduces Aβ loads and Congo Red staining in APP+PS1 transgenic mice

Biochem/physiol Actions

Primary Target
Cyclooxygenase
Target IC50: 5 nM for LPS-induced COX

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 6 months at 4°C. Further dilution of stock solution in aqueous medium should be made just prior to use.

Other Notes

Jantzen, P.T., et al. 2002. J. Neurosci.22, 2246.
Hinz, R., et al. 2001. Pharmacol. Res.18, 151.
Klabunde, T., et al. 2000. Nat. Struct. Biol.7, 312.
Range, S.P., et al. 2000. Eur. Respir. J.15, 751.
van Haeringen, N.J., et al. 2000. J. Ocul. Pharmacol. Ther.16, 353.
Curnock, A.P., et al. 1997. J. Pharmacol. Exp. Ther.282, 339.
Reindeau, D., et al. 1997. Can. J. Physiol. Pharmacol.75, 1088.
Wechter, W.J., et al. 1997. Cancer Res. 57, 4316.
Karmali, R.A., and Marsh, J. 1986. Prostaglandins Leukot. Med. 23, 11.
Karmali, R.A., and Marsh, J. 1985. Prostaglandins Leukot. Med. 20, 283.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service