Skip to Content
Merck
All Photos(1)

Key Documents

329815

Sigma-Aldrich

Erythromycin, Streptomyces erythreus

An antibacterial agent. Inhibits bacterial protein synthesis by binding to the 23S RNA in 50S ribosome.

Synonym(s):

Erythromycin, Streptomyces erythreus, Erythromycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥85% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

ethanol: soluble
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1

InChI key

ULGZDMOVFRHVEP-RWJQBGPGSA-N

General description

An antibacterial agent. Inhibits bacterial protein synthesis by binding to the 23S RNA in 50S ribosome.

Biochem/physiol Actions

Primary Target
bacteria; protein synthesis

Warning

Toxicity: Standard Handling (A)

Other Notes

Weisblum, B. 1984. Br. Med. Bull.40, 47.
Teraoka, H. 1977. Umsch. Wiss. Tech.77, 347.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service