Skip to Content
Merck
All Photos(2)

Key Documents

23-032-M

Sigma-Aldrich

MDM2/CK1δ Protein, active, 10 µg

Active, full length, recombinant human MDM2 (N-terminal GST-tagged) combined in vitro with CK1δ, active (Item # 14-520). For use in Enzyme Assays. Functions as an E3 ligase in ubiquitination assays.

Synonym(s):

Double minute 2 protein, p53-binding protein Mdm2, Hdm2

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
£3,300.00

£3,300.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
£3,300.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

£3,300.00


Please contact Customer Service for Availability

biological source

human

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

GenBank accession no.

General description

MDM2 is an E3 ubiquitin ligase that regulates the turnover of several cellular factors including the p53 tumour suppressor protein and notably MDM2 itself. The suppression of p53 activity by an amplified Mdm2 gene has been observed in many tumours, including soft tissue sarcomas, osteosarcomas and breast tumours.
N-terminal GST-tagged, recombinant human TRAF6 full length, expressed in E.coli.

Application

Ubiquitination Cascade Comoponent: E3

Physical form

Purified using glutathione sepharose followed by gel filtration.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service