Skip to Content
Merck
All Photos(1)

Key Documents

219504

Millipore

Cefotaxime sodium salt

≥97% (HPLC), suitable for cell culture

Synonym(s):

(6 R,7 R)-3-Acetoxymethyl-7-(( Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Na, Cefotaxim sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
Beilstein:
5711411
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

technique(s)

cell culture | mammalian: suitable

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[H][C@]12SCC(COC(C)=O)=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\c3csc(N)n3)C([O-])=O

InChI

1S/C16H17N5O7S2.Na/c1-6(22)28-3-7-4-29-14-10(13(24)21(14)11(7)15(25)26)19-12(23)9(20-27-2)8-5-30-16(17)18-8;/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,23)(H,25,26);/q;+1/p-1/b20-9-;/t10-,14-;/m1./s1

InChI key

AZZMGZXNTDTSME-JUZDKLSSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potent β-lactamase-resistant antibiotic of cephalosporin class. Active against Gram-positive and Gram-negative organisms, including Gram-negative anaerobes. Inhibits cell wall synthesis.

Biochem/physiol Actions

Cefotaxim inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs) which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes.
Mode of Action: Inhibits bacterial cell wall synthesis.
Primary Target
Gram-positive
Secondary Target
Gram-negative

Packaging

Packaged under inert gas

Other Notes

Broad spectrum third generation cephalosporin antibiotic.
Molinari, G., et al. 1991. J. Chemotherap.3, 6.
Vallee, F., and LeBel, M. 1991. Antimicrob. Agents Chemother.35, 2057.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service