Skip to Content
Merck
All Photos(1)

Key Documents

218807

Sigma-Aldrich

Caspase-9, Human, Recombinant, E. coli

Caspase-9, Human, Recombinant is expressed in E. coli as a single polypeptide. Fused at the N- and C- termini to a His•Tag sequence. Undergoes spontaneous autoprocessing to yield the active enzyme.

Synonym(s):

ICE-LAP6, Mch6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in E. coli

Quality Level

Assay

≥70% (SDS-PAGE)

form

lyophilized

specific activity

≥400 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

PBS with 15% glycerol: 1 units/μL

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human caspase-9 fused at the N- and C- termini to a His•Tag sequence and expressed in E. coli as a single polypeptide. Subsequently undergoes spontaneous autoprocessing to yield the active enzyme. MW: 11000 and 20000.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1.0 nmol substrate Ac-LEHD-pNA (Cat. No. 218805) per h at 37°C, pH 7.2.

Physical form

Lyophilized from 130 mM NaCl, 20 mM Tris, 4 mM (NH₄)₂SO₄

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Qin, H., et al. 1999. Nature399, 549.
Li, P., et al. 1997. Cell91, 479.
Salvesen, G.S., et al. 1997. Cell91, 443.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service