Skip to Content
Merck
All Photos(1)

Key Documents

205528

Millipore

C12E8

Constituent of ATLAS G2127 and LUBROL PX. Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state. Aggregation number: 123; HLB number: 13.1.

Synonym(s):

C12E8, Octaethyleneglycol Mono- n-dodecyl Ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H58O9
CAS Number:
Molecular Weight:
538.75
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (GC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

123

CMC

110 μM

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C28H58O9/c1-2-3-4-5-6-7-8-9-10-11-13-30-15-17-32-19-21-34-23-25-36-27-28-37-26-24-35-22-20-33-18-16-31-14-12-29/h29H,2-28H2,1H3

InChI key

YYELLDKEOUKVIQ-UHFFFAOYSA-N

General description

Constituent of ATLAS G2127 and LUBROL PX. Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state. Aggregation number: 123; HLB number: 13.1.
Constituent of ATLAS G2127 and LUBROL PX. Non-ionic detergent useful for the solubilization of membrane-bound proteins in their native state (aggregation number 123; CMC 110 µM; HLB number: 13.1).

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.

Other Notes

Levy, D., et al. 1990. Biochemistry29, 9480.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service