Skip to Content
Merck
All Photos(1)

Key Documents

20-209

Millipore

Catch and Release Non-denaturing Elution Buffer, 4X

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
£80.00

£80.00


Please contact Customer Service for Availability


Select a Size

Change View
10 ML
£80.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.32

£80.00


Please contact Customer Service for Availability

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

Application

Designed for use with the CATCH AND RELEASE reversible immunoprecipitation system to recover captured immunocomplex from CATCH AND RELEASE Spin Columns
Research Category
All

Quality

routinely evaluated as elution buffer for CATCH AND RELEASE (Cat#17-500)

Physical form

PBS based IP Elution Buffer

Storage and Stability

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service