Skip to Content
Merck
All Photos(1)

Documents

196872

Sigma-Aldrich

BAY 11-7085 - CAS 196309-76-9 - Calbiochem

Synonym(s):

(E)3-[(4-t-Butylphenyl)sulfonyl]-2-propenenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥95% (HPLC)

Quality Level

form

solid

color

white

solubility

DMSO: 10 mg/mL

General description

Exhibits biological properties similar to that of BAY 11-7082 (Cat. No. 196870). BAY 11-7085 has also been shown to have potent anti-inflammatory properties in vivo.

Biochem/physiol Actions

  • TNF-α-inducible phosphorylation of I?Bα
  • Target IC<SUB>50</SUB>:10 μM against TNF-α-inducible phosphorylation of I?Bα

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ni et al.
British journal of haematology, 115(2), 279-286 (2001-11-13)
Nuclear factor-kappa B (NF-kappa B) is an important transcription factor that regulates survival in many cells. Activated NF-kappa B has been shown to protect some haematopoietic neoplastic cells from apoptosis. In the present study, we analysed NF-kappa B status in
J W Pierce et al.
The Journal of biological chemistry, 272(34), 21096-21103 (1997-08-22)
We have identified two compounds that inhibit the expression of endothelial-leukocyte adhesion molecules intercellular adhesion molecule-1, vascular cell adhesion molecule-1, and E-selectin. These compounds act by inhibiting tumor necrosis factor-alpha-induced phosphorylation of IkappaB-alpha, resulting in decreased nuclear factor-kappaB and decreased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service