Skip to Content
Merck
All Photos(1)

Key Documents

178220

Sigma-Aldrich

Antipain, Hydrochloride

Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes.

Synonym(s):

Antipain, Hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£64.50
10 MG
£97.90

£64.50


Estimated to ship on28 April 2025Details



Select a Size

Change View
5 MG
£64.50
10 MG
£97.90

About This Item

Empirical Formula (Hill Notation):
C27H44N10O6 · xHCl
CAS Number:
Molecular Weight:
604.70 (free base basis)
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

£64.50


Estimated to ship on28 April 2025Details


Quality Level

description

RTECS - YV9350800

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to slightly yellowish

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].N([C@@H](C(C)C)C(=O)NC(=O)[C@@H](NC(=O)NC(Cc1ccccc1)C(=O)O)CCCN=C(N)N)C(CCCN=C(N)N)C=O.[H+].[H+]

InChI

1S/C27H44N10O6.2ClH/c1-16(2)21(34-18(15-38)10-6-12-32-25(28)29)23(40)37-22(39)19(11-7-13-33-26(30)31)35-27(43)36-20(24(41)42)14-17-8-4-3-5-9-17;;/h3-5,8-9,15-16,18-21,34H,6-7,10-14H2,1-2H3,(H,41,42)(H4,28,29,32)(H4,30,31,33)(H2,35,36,43)(H,37,39,40);2*1H/t18?,19-,20?,21-;;/m0../s1

InChI key

GXQPAKIVOCLCHD-BXLPLHKWSA-N

General description

Effective concentration: 1-100 µM.
Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes. Inhibitor of Ca2+-dependent endopeptidases. Has specificity similar to Leupeptin (Cat. No. 108975). Inhibits trypsin-like serine proteases, papain and some cysteine proteases (IC50 = 300 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Trypsin-like serine proteases
Product does not compete with ATP.
Reversible: no
Target IC50: 300 µM against trypsin-like serine proteases, papain and some cysteine proteases

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 1 week at 4°C or for up to 1 month at -20°C.

Other Notes

Lyupina, Y.V., et al. 1996. Eur. J. Pharmacol. 304, 23.
Umezawa, H. 1976. Methods Enzymol.55, 678.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service